Learn More
MSA™ ForceField™ ACH Accessories and Replacement Parts (Commercial)
Catalog No. 191500361 Shop All MSA Products
Change view
Click to view available options
Size:
6
8
Universal
Quantity:
1 Ea.
1 Kit
10 Kits
Product Type:
Chinstrap Assembly
Pad Kit
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Quantity Product Type
19-150-0361 8 10 Kits Pad Kit
19-150-0358 6 1 Kit Pad Kit
19-150-0359 8 1 Kit Pad Kit
19-150-0357 Universal 1 Ea. Chinstrap Assembly
19-150-0360 6 10 Kits Pad Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-150-0361 Supplier MSA™ Supplier No. 10060736
For use with MSA ForceField ACH Advanced Combat Helmet
Compliance
  • Ballistic resistance: Meets NIJ 0108.01 IIIa standards
  • Fragment/impact resistance: Meets Military ACH specifications
Specifications
Certifications/Compliance NIJ 0108.01 IIIa
For Use With (Equipment) ACH Advanced Combat Helmet
Product Type Pad Kit
Quantity 10 Kits
Size 8