Learn More
MSA™ ForceField™ Concealable Body Armor Accessories: Trauma Plates
Catalog No. 191500383 Shop All MSA Products
This item is not returnable. View return policy
Catalog No. 19-150-0383 Supplier MSA™ Supplier No. 10067348
This item is not returnable. View return policy
For MSA ForceField Concealable Body Armor
Soft Ballistic Trauma Pack
  • 100% woven Kevlar™ with nylon urethane coated cover and durable water resistant finish
  • Fit ForceField Alpha or Bravo Concealable Vest
  • Dimensions: 6 x 8 in. (15.25 x 20.25cm)
Compliance
Meet NIJ 0101.04 standards.
Specifications
Product Type Trauma Plates
Material Woven Dupont™ Kevlar™ aramid fiber, Nylon Urethane
Certifications/Compliance NIJ 0101.04