Learn More
MSA™ Goggle Retainers
Numéro de catalogue. 19041447 missing translation for 'brandsLinkText'
Modifier l'affichage
Click to view available options
For Use With (Equipment):
For MSA Caps
For MSA Hats
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. For Use With (Equipment)
19-041447 For MSA Caps
18-999-3895 For MSA Hats
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 19-041447 Fournisseur MSA™ Code fournisseur 459458
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Holds goggles securely on any MSA cap or hat
  • Sturdy neoprene-rubber accessory can be used with any MSA protective cap or hat
  • Retains goggle on helmet when not in use and holds goggle securely when worn for eye protection
Compliance
MSA goggle retainers are manufactured for use ONLY with MSA goggles in order to comply with ANSI/ISEA Z87.1-2010.
Spécifications
Product Type Goggle Retainer
For Use With (Equipment) For MSA Caps
For Use With (Application) Make goggles conveniently available for immediate use
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.