Learn More
MSA™ Pulley and Bracket Assembly
Catalog No. 19818270 Shop All MSA Products
Change view
Click to view available options
Side-Mount Pulley with Bracket
Catalog No.
19-818-270 Side-Mount Pulley with Bracket
1 options
Catalog No. 19-818-270 Supplier MSA™ Supplier No. 506473
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
  • Features split-mount that can be used with tripod or remotely anchored to a suitable anchorage point makeing the device highly portable
  • Side-Mount Dyna-Hoist™ is designed for lifting, lowering and positioning of personnel and materials with a maximum load of 310 lb.
  • Line comes with 50 ft. of 3/16 in. galvanized rope
  • Meets or exceeds applicable ANSI and OSHA requirements
Specifications
Product Type Pulley and Bracket Assembly
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.