Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Lens Cover for Ultra Elite Facepiece

Catalog No. 189993626 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
Ultra elite full facepiece respirator
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-999-3626 Ultra elite full facepiece respirator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-999-3626 Supplier MSA™ Supplier No. 491500
Add to Cart
Add to Cart

Clear overlay lens cover accessory is available 25/pk.

  • Accessory for Ultra Elite facepiece
  • 25/pack

Specifications

Color Clear
Product Type Lens Cover
For Use With (Equipment) Ultra elite full facepiece respirator
Model 491500
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.