Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Millennium™ CBRN Gas Mask

Catalog No. 191300460 Shop All MSA Products
Change view
Click to view available options
Format:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
19-130-0460 Large
19-130-0458 Small
19-130-0459 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-130-0460 Supplier MSA™ Supplier No. 10051288
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-performance, Hycar™ rubber gas mask is ideal for use by law enforcement or emergency response personnel

Based on the design of the MCU-2/P mask used by the U.S. Air Force in Operation Desert Storm, this CBRN-approved gas mask accommodates a military-style, 40mm threaded canister. Dual-canister mounts permit weapon sighting from either shoulder. Super-soft, lightweight facepiece construction contributes to a snug, comfortable fit. Flexible lens is bonded to the chemical-resistant Hycar facepiece as part of a wraparound design that minimizes wasted space and offers a wide field of vision.

  • Fully elastic, six-point head harness that permits easy donning, doffing and adjustment without pulling hair
  • Speaking diaphragm for ease of communications
  • Drinking tube that can be used for replenishing fluids
  • Nosecup with two check valves that deflects exhaled air away from the one-piece polyurethane lens, preventing fogging
  • Snap-in polycarbonate lens outsert for extra protection of the lens surface

Order Info

Overall size is compatible with most visored police helmets.

Specifications

Certifications/Compliance NIOSH, CBRN
Connection Type 40 mm thread
Material Hycar Rubber
Product Type Millennium™ Cbrn Gas Mask
Format Large
For Use With (Application) CBRN incidents, civil disturbances, clandestine labs
Includes Drinking port, speaking diaphragm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.