Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Millennium™ Gas Mask Lens Outserts

Catalog No. 191300548 Shop All MSA Products
Change view
Click to view available options
Size:
Medium/Large
Small
Color:
Clear
Tinted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19-130-0548 Medium/Large Tinted
19-130-0545 Small Clear
19-130-0546 Medium/Large Clear
19-130-0547 Small Tinted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-130-0548 Supplier MSA™ Supplier No. 10008908
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polycarbonate lens outsert snaps into place over Millennium or Advantage 1000 facepiece lens, providing additional impact protection.

  • Accessory for Millennium or Advantage 1000 facepieces
  • Polycarbonate outsert
  • Available in clear or tinted
  • Available in small or medium/large sizes

Specifications

Color Tinted
Material Polycarbonate
Product Type Lens Outsert
Size Medium/Large
For Use With (Application) Provide additional impact protection
For Use With (Equipment) Millennium™ or Advantage 1000 facepiece
Model 10008908
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.