Learn More
MSA™ Model R Calibration Cylinder Holder
Catalog No. 189993516 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
Model R Calibration Cylinder
Catalog No. For Use With (Equipment)
18-999-3516 Model R Calibration Cylinder
1 options
Catalog No. 18-999-3516 Supplier MSA™ Supplier No. 710483
Add to Cart
Add to Cart
Holder for cylinders. Complete with securing straps and sized to house Model R, RP and Econo-Cal Cylinders. Note: Photo at left shows holder with cylinders (not included).
  • Wall- or bench-mount allows user to house two cylinders, with or without regulators attached
  • Molded base locates cylinders in position
  • Easy to load and unload cylinder
Specifications
For Use With (Equipment) Model R Calibration Cylinder
Product Type Dual Cylinder Holder