Learn More
MSA™ Model RP Combination Regulators
Catalog No. 18039209 Shop All MSA Products
Change view
Click to view available options
Flow Rate:
0.25 L/min.
1.5 L/min.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate
18-039-209 0.25 L/min.
18-039-210 1.5 L/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-039-209 Supplier MSA™ Supplier No. 711175
Add to Cart
Add to Cart
A fixed flow design provides constant and accurate flow to the gas detector simplifying the test and calibration process and saving calibration gas
  • Two regulators in one, designed for use with all Model RP and Econo-Cal calibration cylinders
  • No ON/OFF valve is necessary, because the regulator automatically turns itself off when not in use
  • Trigger activation allows a bump test or calibration check in the "SQUEEZE" mode or fixed-flow calibration in the up or "LOCKED" position
Specifications
Flow Rate 0.25 L/min.
For Use With (Equipment) Model RP
Product Type Respirator Assembly
For Use With (Application) Agriculture, chemical, construction, electric utility, fire fighting, with calibration tubing
Includes Calibration tubing