Description
- Chemical components: Cadmium: 11-26%, Nickel: 8-17%, Potassium Hydroxide: <3%, Nylon: <2%, Steel: 12-13%, Lead as solder: Trace and others (plastic): <10%
- State of charge and environment have impact on contents of battery
- According to 29 CFR 1910.1200 (c) defined as article
- Exemption from the U.S. OSHA Hazard Communication Standard
Specifications
Specifications
| Product Type | NiCd Battery Pack |
| For Use With (Equipment) | FiveStar alarm |