Learn More
MSA™ Ultra Elite Full Facepiece
Catalog No. 189993164 Shop All MSA Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
18-999-3164 Black
1 options
This item is not returnable. View return policy
Catalog No. 18-999-3164 Supplier MSA™ Supplier No. 805331
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ultra Elite full-face mask features a comfortable ergonomic design, while the 5-point harness makes it easy to put it on or take it off.
  • Transmitter-receiver assembly for monitoring air cylinder contents
  • Designed for Ultra Elite facepiece
  • Features low-battery indicator and 7 LED light patterns
  • Available in three sizes
Specifications
Certifications/Compliance NIOSH
Color Black
Connection Type Pressure-Demand
Product Type Facepiece Assembly
Includes Nosecup, Speed-On Head Harness
Material BMR, Hycar
Size Large
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.