Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Airline Respirator Accessory, Nipple Pipe

Catalog No. 18139726 Shop All MSA Products
Catalog No. 18-139-726 Supplier MSA™ Supplier No. 68833
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Connects air-supply hoses to manifolds and to air source.

Specifications

Model 68833
Overall Length (English) 1.5 in.
Overall Length (Metric) 3.8 cm
Overall Width (English) 0.75 in.
Overall Width (Metric) 1.9 cm
Overall Height (English) 0.75 in.
Overall Height (Metric) 1.9 cm
Product Type Pipe
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.