Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Nosecups for Ultravue™ and Ultra-Twin™ Facepieces

Catalog No. 19037292 Shop All MSA Products
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-037-292 Medium
18-999-3654 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-037-292 Supplier MSA™ Supplier No. 813139
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Black silicone replacement nosecups for use with Ultravue and Ultra-Twin facepieces.

  • Replacement black silicone nosecup
  • Easily affixed to the facepiece without any tools
  • Reduces the possibility of lens fogging
  • Available in medium or large sizes

Specifications

Color Black
Material Silicone
Product Type Nosecup Assembly
Size Medium
For Use With (Application) Helps reduce lens fogging
For Use With (Equipment) Ultravue and Ultra-Twin Full Facepieces
Model 813139
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.