Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ OptimAir™ Mask-Mounted PAPR Accessory, Elite Carrying Case

Catalog No. 19037722 Shop All MSA Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-037-722 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-037-722 Supplier MSA™ Supplier No. D2056734
Add to Cart
Add to Cart

Carrying and storage case for use with OptimAir facemask.

Specifications

Product Type Carrying Case
Color Black
Material Plastic
Model D2056734
For Use With (Equipment) OptimAir Mask-Mounted PAPR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.