Learn More
MSA OptimAir™ TL PAPR Facepiece
Catalog No. 19716752 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
TL powered air purifying respirator
Catalog No. For Use With (Equipment)
19-716-752 TL powered air purifying respirator
1 options
Catalog No. 19-716-752 Supplier MSA Supplier No. 10083800
Add to Cart
Add to Cart
For use with OptimAir TL PAPR
APF of 1000 for tight-fitting facepieces.
Advantage™ 4000 Facepieces
  • Single-port with nosecup; no adapter
  • Silicone with cloth head harness
  • Fit a wide range of face sizes
Specifications
Certifications/Compliance NIOSH
Material Silicone
Product Type Facepiece Assembly
Size Large
For Use With (Application) Asbestos abatement, welding, sanding, grinding
For Use With (Equipment) TL powered air purifying respirator
Includes Motor blower, smart charger, standard battery, hose and decon belt, standard connectors and suspension
Model 10083800