Learn More
MSA™ OptimAir™ TL PAPR: High-Efficiency Cartridges
Catalog No. 18808765 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
TL powered air purifying respirator
Catalog No. For Use With (Equipment)
18-808-765 TL powered air purifying respirator
1 options
Catalog No. 18-808-765 Supplier MSA™ Supplier No. 10080455
Add to Cart
Add to Cart
For use with OptimAir TL PAPR
Certifications
NIOSH approved.
Specifications
Certifications/Compliance NIOSH
Filter Type P100
Product Type HE Cartridge
For Use With (Application) Welding, grinding and sanding
For Use With (Equipment) TL powered air purifying respirator
Weight (English) 2.5 lb.
Weight (Metric) 1.133 kg