Learn More
MSA OptimAir™ TL PAPR: Single Bib Hoods
Catalog No. 19716746 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
TL powered air purifying respirator
Catalog No. For Use With (Equipment)
19-716-746 TL powered air purifying respirator
1 options
Catalog No. 19-716-746 Supplier MSA Supplier No. 10083383
Add to Cart
Add to Cart
For use with OptimAir TL PAPR
APF of 1000 for hoods.
Tychem™ Hoods
  • Available in polyethylene-coated Tyvek™ (QC) or Saranex-coated Tyvek (SL) with single or double bib
  • Can be worn with a hardhat for protection against overhead hazards
  • Oversized, optically improved lens for greater field of vision with less distortion
Specifications
Certifications/Compliance NIOSH
Material Tyvek
Product Type Single Bib Hood
For Use With (Application) Welding, grinding and sanding
For Use With (Equipment) TL powered air purifying respirator
Includes Motor blower, smart charger, standard battery, hose and decon belt, standard connectors and suspension
Model 10083383