Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ OptimAir™ TL Powered Respirator Combination Cartridges

Catalog No. 18808766 Shop All MSA Products
Change view
Click to view available options
Cartridge Type:
AG, AM, FM, HE, OV
AG, HEPA, HF, OV
Color:
Magenta/Olive
Magenta/Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cartridge Type Color
18-808-766 AG, AM, FM, HE, OV Magenta/Olive
18-808-764 AG, HEPA, HF, OV Magenta/Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-808-766 Supplier MSA™ Supplier No. 10080456
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vapor cartridges install on PAPR systems to filter contaminants from ambient air.

  • Quantity of 6 per pack
  • Choose from two options:
    • 10080454 Combination cartridge effective against OV/AG/HE
    • 10080456 Combination cartridge effective against AG/AM/FM/HE/OV
  • NIOSH-approved
  • Bayonet style connection

Specifications

Cartridge Type AG, AM, FM, HE, OV
Color Magenta/Olive
Product Type Cartridge
For Use With (Equipment) OptimAir™ TL PAPR Systems
Quantity 6/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.