Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Phalanx™ CBA-RCA Gas Mask Canister

Catalog No. 19500320 Shop All MSA Products
Change view
Click to view available options
Canister; box of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-500-320 Canister; box of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-500-320 Supplier MSA™ Supplier No. 817591

Phalanx canisters for use with CBA-RCA (Chemical Biological Agent- Riot Control Agent) gas masks.

  • NIOSH-approved for protection against:
    • Chloroacetophenone (CN)
    • Chlorobenzylidene (CS)
    • P100 particulate efficiency level
    • Particulates
  • Gas mask canisters filter airborne contaminants for CBRN (chemical, biological, radiological, and nuclear) environments
  • Designed to protect against one or more specific hazards and are used with compatible headgear
  • Canisters can be dual-mounted on the gas mask to allow weapon firing from either shoulder

Specifications

Color Black
Product Type CBRN Canister
For Use With (Application) CBRN incidents, civil disturbances, clandestine labs
For Use With (Equipment) Phalanx cba-rca gas mask
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.