Learn More
MSA™ PremAire™ Cadet Escape Respirator Kits
Catalog No. 17200032 Shop All MSA Products
Change view
Click to view available options
Includes:
Airline respirator, advantage™ 4000 facepiece with push-to-connect firehawk™ mmr nylon belt assembly, 10 min. aluminum egress cylinder, hansen brass quick-disconnects, 50 ft. section of neoprene air-supply hose with han
Airline respirator, advantage™ 4000 facepiece with push-to-connect firehawk™ mmr nylon belt assembly, 5 min. aluminum egress cylinder, hansen brass quick-disconnects, 50 ft. section of neoprene air-supply hose with hans
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
17-200-032 Airline respirator, advantage™ 4000 facepiece with push-to-connect firehawk™ mmr nylon belt assembly, 10 min. aluminum egress cylinder, hansen brass quick-disconnects, 50 ft. section of neoprene air-supply hose with han
17-200-031 Airline respirator, advantage™ 4000 facepiece with push-to-connect firehawk™ mmr nylon belt assembly, 5 min. aluminum egress cylinder, hansen brass quick-disconnects, 50 ft. section of neoprene air-supply hose with hans
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-200-032 Supplier MSA™ Supplier No. 10108592
Add to Cart
Add to Cart
Includes
  • Combination supplied-air PremAire Cadet escape respirator with egress cylinder
  • Advantage™ 4000 facepiece (medium size, Hycar™ rubber, polyester net head harness) with Push-to-Connect FireHawk MMR nylon belt assembly
  • 50 ft. section of neoprene air-supply hose with Hansen brass plug
  • Hansen brass quick disconnects
Low-profile escape cylinders may be worn on either hip for user comfort
Specifications
Certifications/Compliance NIOSH
Product Type Cadet Escape Respirator
For Use With (Application) Welding
For Use With (Equipment) Premaire™ cadet escape respirator
Includes Airline respirator, advantage™ 4000 facepiece with push-to-connect firehawk™ mmr nylon belt assembly, 10 min. aluminum egress cylinder, hansen brass quick-disconnects, 50 ft. section of neoprene air-supply hose with han
Quantity 1 Ea.