Learn More
MSA™ PremAire™ Cadet Escape Respirators
Catalog No. 17200030 Shop All MSA Products
Change view
Click to view available options
Includes:
10 min. aluminum 3000 psi cylinder, nylon strap harness/carrier, first stage regulator and cylinder valve
10 min. aluminum 3000 psi cylinder, nylon strap harness/carrier, ultra elite hycar™ facepiece with rubber head harness, hard case, first stage regulator and cylinder valve
5 min. aluminum 2216psi cylinder, nylon strap harness/carrier, advantage™ 4000 hycar™ facepiece with rubber head harness, hard case, first stage regulator and cylinder valve
5 min. aluminum 2216psi cylinder, nylon strap harness/carrier, first stage regulator and cylinder valve
5 min. aluminum 2216psi cylinder, nylon strap harness/carrier, ultra elite hycar™ facepiece with rubber head harness, hard case, first stage regulator and cylinder valve
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
17-200-030 5 min. aluminum 2216psi cylinder, nylon strap harness/carrier, advantage™ 4000 hycar™ facepiece with rubber head harness, hard case, first stage regulator and cylinder valve
17-200-026 5 min. aluminum 2216psi cylinder, nylon strap harness/carrier, first stage regulator and cylinder valve
17-200-027 10 min. aluminum 3000 psi cylinder, nylon strap harness/carrier, first stage regulator and cylinder valve
17-200-028 5 min. aluminum 2216psi cylinder, nylon strap harness/carrier, ultra elite hycar™ facepiece with rubber head harness, hard case, first stage regulator and cylinder valve
17-200-029 10 min. aluminum 3000 psi cylinder, nylon strap harness/carrier, ultra elite hycar™ facepiece with rubber head harness, hard case, first stage regulator and cylinder valve
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-200-030 Supplier MSA™ Supplier No. 10093988
Add to Cart
Add to Cart
Streamlined design with a first-stage regulator and cylinder within one assembly
  • Low-profile escape cylinder can be worn on either hip for user comfort
  • Small size is less likely to snag when working in tight spaces
  • Two attachments for keeping cylinder secure on belt
  • Shoulder support strap incorporates hose keeper; regulator hose stays in place close to body
  • Bracket/holder of second stage pressure regulator
  • Nylon strap harness/carrier
Certifications
NIOSH approved as open-circuit, pressure-demand escape SCBA, open-circuit, pressure-demand escape SCBA and Type C supplied-air respirator
Compliance
NIOSH approved as open circuit; pressure-demand escape SCBA; open-circuit, pressure-demand escape SCBA; and Type C supplied-air respirator.
Specifications
Certifications/Compliance NIOSH
Product Type Cadet Escape Respirator
For Use With (Application) Welding, chemical/petrochemical, hazardous materials, confined space entry, firefighting operation
Includes 5 min. aluminum 2216psi cylinder, nylon strap harness/carrier, advantage™ 4000 hycar™ facepiece with rubber head harness, hard case, first stage regulator and cylinder valve
Quantity 1 Ea.