Learn More
MSA™ PremAire™ Cadet Respirator Kit
Catalog No. 17200023 Shop All MSA Products
Change view
Click to view available options
Connection Type:
Foster steel quick-disconnect
Snap-tite™ aluminum quick-disconnect
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
17-200-023 Foster steel quick-disconnect
17-200-025 Snap-tite™ aluminum quick-disconnect
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-200-023 Supplier MSA™ Supplier No. 10108493
Add to Cart
Add to Cart
In a non-IDHL environment, the MSA PremAir Cadet Respirator Kit is an ideal choice for protecting workers in applications such as painting, asbestos abatement, sanding, wastewater treatment, food processing and more.
For use in non-IDLH (immediately dangerous to life or health) environments only.
  • Pressure-demand, Type C, NIOSH-approved supplied-air PremAire Cadet respirator
  • Advantage™ 4000 facepiece (medium size, Hycar™ rubber, polyester net head harness) with Push-to-Connect FireHawk MMR nylon belt assembly
  • 50 ft. section of neoprene air-supply hose with plug (corresponds to air source)
  • Choice of Foster steel or Snap-Tite aluminum quick-disconnect
Order Info
Additional respirators available separately (See Cat. No. 17-200-024).
Specifications
Certifications/Compliance NIOSH
Connection Type Foster steel quick-disconnect
Product Type Supplied Air Respirator Kit
For Use With (Application) Asbestos abatement, painting, sanding, grinding, welding
For Use With (Equipment) PremAire™ Cadet Respirator
Model 10108493