Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Pressure Demand Air-Line Respirators

Catalog No. 189993109 Shop All MSA Products
Change view
Click to view available options
Material:
Foster Steel
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
18-999-3109 Stainless Steel
18-999-3115 Foster Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-999-3109 Supplier MSA™ Supplier No. 475221

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Full-facepiece respirators feature belt-mounted regulator that maintains slight positive pressure of air inside facepiece during inhalation and exhalation to prevent influx of contaminants. Assembly includes regulator and quick-disconnect, breathing tube, Ultravue™ facepiece, and web belt. Facepiece features scratch-resistant polycarbonate lens with wraparound design for minimal visual distortion, inhalation check valve that prevents exhaled air from entering breathing tube or regulator, mechanical speaking diaphragm for short-range communications, and five adjustable rubber straps and a vinyl neck strap for holding facepiece in standby position.
Respirators are designed for use in non-IDLH atmospheres and are suitable for applications such as welding and painting, and for handling certain hazardous materials. Available with stainless-steel or Foster steel quick-disconnect.

Compliance

NIOSH/MSHA approved for operation at 65 to 85psig using 8 to 300 ft. (2.4 to 91.4m) of MSA airline hose.

Specifications

Certifications/Compliance NIOSH, MSHA
Material Stainless Steel
Product Type Quick-Disconnect
For Use With (Application) Non-IDLH atmospheres, welding and painting and for handling certain hazardous materials
Includes Regulator and quick-disconnect, breathing tube, ultravue™ facepiece and web belt
Material (Lens) Polycarbonate
Pressure 65 to 85 psi
Resistant To Scratch-resistant
Length (English) Hose 8 to 300 ft.
Length (Metric) Hose 2.43 to 91.44 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.