Learn More
MSA™ Air Pressure Regulator
Catalog No. 189993219 Shop All MSA Products
Change view
Click to view available options
Pressure demand regulator
Catalog No.
18-999-3219 Pressure demand regulator
1 options
Catalog No. 18-999-3219 Supplier MSA™ Supplier No. 470193
Constant flow airline accessory.
  • Pressure regulator reduces plant air system compressor pressure to desired operating pressure for respirators being used
  • Maintains pressure on outlet side until re-adjusted for use with inlet pressures of up to 125 psig
Specifications
For Use With (Equipment) Airline Respirator
Model 470193
Product Type Pressure Demand Regulator