Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Probed Respirator Facepieces

Catalog No. 189993691 Shop All MSA Products
Change view
Click to view available options
Size:
Medium
Large
Color:
Black
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
18-999-3691 Large Black
18-999-3690 Medium Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-999-3691 Supplier MSA™ Supplier No. 808259
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Product Type Probed Respirator Facepiece
Color Black
For Use With (Application) Respirator Fit Testing
Includes Built-in probe assembly, solid brass probe and brass fitting
Model 808259
Size Large
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.