Learn More
MSA™ ProCheck™ III Software
Catalog No. 19815414 Shop All MSA Products
Change view
Click to view available options
PosiChek Software Cd
Catalog No.
19-815-414 PosiChek Software Cd
1 options
Catalog No. 19-815-414 Supplier MSA™ Supplier No. 10006002
Requires
Biosystems PosiChek III Computerized SCBA Performance Tester
Walks technician through step-by-step inspection process
Inspects any MSA air mask or supplied-air respirator
Specifications
Product Type Procheck Iii Software Cd
For Use With (Application) Respirator Fit Testing
For Use With (Equipment) Biosystems procheck™ iii computerized scba performance tester
Model 10006002
Quantity 1 Ea.