Learn More
MSA™ Quantitative Fit Test Adapter
Catalog No. 19716757 Shop All MSA Products
Change view
Click to view available options
Certifications/Compliance:
NOSH
Catalog No. Certifications/Compliance
19-716-757 NOSH
1 options
Catalog No. 19-716-757 Supplier MSA™ Supplier No. 10056424
Specifications
Product Type Fit Test Adapter
Certifications/Compliance NOSH
For Use With (Application) Allows quantitative fit tests to be performed on bayonet cartridge style respirators
For Use With (Equipment) Advantage™ 200ls half mask and advantage 3200 full facepiece respirators
Model 10056424
Quantity 1 Ea.
Weight (English) 0.20 lb.
Weight (Metric) 0.9 kg