Learn More
MSA™ Quick Disconnect Accessories for Airline Respirators and Hoods
Catalog No. 19120443 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
Air hose
Catalog No. For Use With (Equipment)
19-120-443 Air hose
1 options
Catalog No. 19-120-443 Supplier MSA™ Supplier No. 69541
Add to Cart
Add to Cart
For use with supplied air respirators.
  • Adapter
  • Union brass
  • 0.25 in. NPT male X 0.75 in. female
  • MSA 69541
Specifications
Material Brass
Product Type Adapter
For Use With (Equipment) Air hose
Model 69541