Learn More
MSA™ Quick-Disconnects and Adapters: Adjustable Valve, Continuous Flow
Catalog No. 19815477 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
MSA Quick-Disconnects and Adapters
Catalog No. For Use With (Equipment)
19-815-477 MSA Quick-Disconnects and Adapters
1 options
Catalog No. 19-815-477 Supplier MSA™ Supplier No. 463265
Add to Cart
Add to Cart
Continuous flow valve for supplied air respirators.
  • Low-pressure control valve
  • Adjustable continuous flow
Specifications
Product Type Continuous Flow Adjustable Valve
For Use With (Equipment) MSA Quick-Disconnects and Adapters
Model 463265