Learn More
MSA™ Air-Line Respirators and Air-Supplied Hoods Quick-Disconnect Accessory, Female Socket Assembly
Catalog No. 19815488 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
Quick-Disconnect
Catalog No. For Use With (Equipment)
19-815-488 Quick-Disconnect
1 options
Catalog No. 19-815-488 Supplier MSA™ Supplier No. 471778
Add to Cart
Add to Cart
Assembly is used to connect hose to male plug on supplied-air respirator.
  • Quick-disconnect accessory
  • Stainless steel
  • Female socket
Specifications
Material Stainless Steel
Product Type Hose Socket
For Use With (Application) Connect air supply hoses to manifold and air source
For Use With (Equipment) Quick-Disconnect
Model 471778