Learn More
MSA™ Air-Line Respirators and Air-Supplied Hood Accessory, Quick-Disconnect Hansen Brass Male Plug
Catalog No. 189994006 Shop All MSA Products
Change view
Click to view available options
Includes:
0.25 in. NPT female
Catalog No. Includes
18-999-4006 0.25 in. NPT female
1 options
Catalog No. 18-999-4006 Supplier MSA™ Supplier No. 630313
Add to Cart
Add to Cart
Assembly is used to connect hose to female plug on supplied-air respirator.
  • Quick-disconnect accessory
  • Stainless steel
  • Male socket
Specifications
Material Brass
Product Type Plug
For Use With (Application) Connect air supply hoses to manifold and air source
For Use With (Equipment) Airline Respirator and Air-Supplied Hood
Includes 0.25 in. NPT female
Model 630313