Learn More
MSA™ Quick-Disconnect Male Plug with Male NPT
Catalog No. 18139737 Shop All MSA Products
Change view
Click to view available options
Connection Type:
Male 0.25 in. NPT
Catalog No. Connection Type
18-139-737 Male 0.25 in. NPT
1 options
Catalog No. 18-139-737 Supplier MSA™ Supplier No. 473501
Add to Cart
Add to Cart
Brass quick-disconnect used for connecting inlet pressure gauge.
Male 1/4 in. NPT
Fittings with the same letter code are interchangeable.
  • AL: Aluminum
  • S: Steel
  • SST: Stainless Steel
  • BR: Brass
Specifications
Connection Type Male 0.25 in. NPT
For Use With (Equipment) Airline Respirator and Air-supplied Hood
Material Brass
Model 473501
Product Type Foster