Learn More
MSA™ Air-Line Respirator / Air-Supplied Hood Accessory, Quick-disconnect Male Plug with Male NPT
Catalog No. 189993968 Shop All MSA Products
Change view
Click to view available options
Snap-Tite; Aluminum
Catalog No.
18-999-3968 Snap-Tite; Aluminum
1 options
Catalog No. 18-999-3968 Supplier MSA™ Supplier No. 066273
For consistent reliability when connecting an inlet pressure gauge, choose an MSA brass or aluminum Quick-disconnect for your system.
Male 1/4 in. NPT
Fittings with the same letter code are interchangeable.
  • AL: Aluminum
  • S: Steel
  • SST: Stainless Steel
  • BR: Brass
Specifications
Connection Type Male 1/4 in. NPT
Material Aluminum
Product Type Hose Assembly
For Use With (Application) Connection of inlet pressure gauge
For Use With (Equipment) Airline respirator and air-supplied hood
Model 066273