Learn More
MSA™ Quick-Fill™ System Accessories
Catalog No. 189993240 Shop All MSA Products
Change view
Click to view available options
Product Type:
Female Coupling
Catalog No. Product Type
18-999-3240 Female Coupling
1 options
Catalog No. 18-999-3240 Supplier MSA™ Supplier No. 485392
Add to Cart
Add to Cart
Quick-disconnect assemblies for use with supplied-air respirators.
Specifications
For Use With (Equipment) Quick-fill System
Includes Sae-4 Thread, Dust Cover
Model 485392
Product Type Female Coupling