Learn More
MSA™ QuickCheck™ III Fit Test Adapter
Catalog No. 19815682 Shop All MSA Products
Change view
Click to view available options
Model:
805078
Catalog No. Model
19-815-682 805078
1 options
Catalog No. 19-815-682 Supplier MSA™ Supplier No. 805078
Add to Cart
Add to Cart
Requires
For completion of a quantitative fit test, an OptiFilter™ XL Cartridge must then be threaded into the QuikCheck III Adapter
Adapter is specially designed for Ultra Elite© Full Facepieces equipped with a coupling nut
  • QuikChek™ Series Adapters attach easily to MSA facepieces
  • Eliminates need for special probed facepieces for quantitative tests
  • Asbestos Abatement
  • Welding
  • Sanding & Grinding
  • Painting
Order Info
Filter not included
Specifications
Product Type Fit Test Adapter
For Use With (Equipment) Ultravue™, ultra elite™ and advantage™ 4100 respirators which are equipped with ultra elite™ full facepieces by attaching to the bmr adapter assembly
Model 805078
Quantity 1 Ea.