Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Ultra Elite™ Positive Pressure Full Facepiece Assemblies

Catalog No. 19005406A Shop All MSA Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-005-406A Small
19-005-406C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-005-406A Supplier MSA™ Supplier No. 10037650
Add to Cart
Add to Cart
This item is not returnable. View return policy

Facepiece features a distortion-free lens that offers wide field-of-view. Ideal for civil disturbances, firefighting, flame detection, and search and rescue. Available in small and large sizes.

  • Ultra Elite Nosecup
  • Hycar material
  • Rubber head harness
  • Non-NFPA
  • Firehawk PTC MMR regulator

Specifications

Certifications/Compliance NIOSH
Color Black
Product Type Full Facepiece Respirator
For Use With (Application) Civil disturbances, firefighting, flame detection, and search and rescue
For Use With (Equipment) Ultra Elite™ Series
Includes Head harness and nosecup
Material Hycar
Size Small

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.