Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Replacement Facepiece Assemblies for Ultra Elite™ and Ultravue™ SCBAs

Catalog No. 19005404B Shop All MSA Products
Change view
Click to view available options
Size:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-005-404B Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-005-404B Supplier MSA™ Supplier No. 10035682
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hycar rubber facepiece is a replacement part for Ultra Elite and Ultravue assemblies.

Replacement part. Size medium. For use with MSA facepieces only. Constructed of Hycar rubber material.

Specifications

Certifications/Compliance NIOSH 42CFR, part 84 NIOSH CBRN NFPA 1981-2002, NFPA 1982-2002
Color Black
Applications Firefighting
For Use With (Equipment) Firehawk MMR Air Mask
Material Hycar Rubber
Includes Speed-on™ head harness, nosecup
Model 10035682
Size Medium
Product Type MMR Facepiece
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.