Learn More
MSA™ Rope Grab
Catalog No. 191600214 Shop All MSA Products
Change view
Click to view available options
Certifications/Compliance:
ANSI Z359.1, OSHA, CSA, N 353-2
Catalog No. Certifications/Compliance
19-160-0214 ANSI Z359.1, OSHA, CSA, N 353-2
1 options
Catalog No. 19-160-0214 Supplier MSA™ Supplier No. 415940
Add to Cart
Add to Cart
Provides safety and ease of mind while working
  • Essential roofing equipment meant to reduce accidents on the job
Certifications
EN 353-2
Compliance
Meets applicable OSHA standards; ANSI™ Z359.1; EN 353-2: CSA 2012-12-12 requirements.
Specifications
Includes 3 ft. Dyna brake shock absorbing lanyard
Certifications/Compliance ANSI Z359.1, OSHA, CSA, N 353-2
Product Type Rope Grab
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.