Learn More
MSA™ Saccharin Qualitative Fit-Test Kits
Catalog No. 189993275 Shop All MSA Products
Change view
Click to view available options
Saccharin fit test kit
Catalog No.
18-999-3275 Saccharin fit test kit
1 options
This item is not returnable. View return policy
Catalog No. 18-999-3275 Supplier MSA™ Supplier No. 697287
This item is not returnable. View return policy
Includes
Two nebulizer bulbs, testing solution insert, sensitivity test solution insert, test hood, one replacement set of testing and sensitivity nebulizer inserts, and Using the Qualitative Fit-Test Kit videotape.
Kit contains everything needed to perform OSHA-compliant qualitative fit tests on disposable and reusable dust/mist respirators
Eliminates the need for special filters.
Specifications
Product Type Saccharin Fit Test Kit
For Use With (Application) Qualitative fit tests on disposable and reusable dust/mist respirators
Includes Two nebulizer bulbs, testing solution insert, sensitivity test solution insert, test hood, one replacement set of testing and sensitivity nebulizer inserts, and Using the Qualitative Fit-Test Kit videotape.
Model 697287
Quantity 1 Ea.