Learn More
Description
- Translucent hood with one-piece large bonded lens offers secure protection and a wide field of view
- Canister is permanently bonded to the respirator for a secure connection
- Rubber neck seal provides a comfortable and reliable seal
- Elastic hair tie allows for the securing of hair before donning the respirator
- Nose cup helps reduce lens fogging and carbon dioxide buildup
- Hexagonal storage case is strong, lightweight, and easily accessible
- 15 minute air supply
Standard
- Color-coded labels on respirator
- Black/gray labels on respirator
Guards against HD vapor, GB, Ammonia, Cyanogen chloride, Cyclohexane, Formaldehyde, Hydrogen cyanide, Hydrogen sulfide, Nitrogen dioxide, Phosgene, Phosphine, sulfur dioxide
Certifications
NIOSH - 10052317
Specifications
Specifications
| Certifications/Compliance | NIOSH |
| Product Type | Standard Escape Respirator |
| Duration | 15 min. |
| Cycle Time | 15 min. |
| For Use With (Application) | CBRN incidents, civil disturbances, clandestine labs |
| Includes | Bonded lens and cbrn canister, high stretch rubber neck seal, nose cup |
| Model | 10066791 |
| Size | Medium |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.