Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Safe Escape CBRN Respirators

Catalog No. 19161116 Shop All MSA Products
Change view
Click to view available options
Product Type:
Standard Escape Respirator
Tactical Escape Respirator
Size:
Small
Medium
Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Size
19-161-116 Tactical Escape Respirator Large
19-161-114 Tactical Escape Respirator Small
19-161-115 Tactical Escape Respirator Medium
19-161-113 Standard Escape Respirator Large
19-161-112 Standard Escape Respirator Medium
19-161-111 Standard Escape Respirator Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-161-116 Supplier MSA™ Supplier No. 10066789

missing translation for 'validMainframeMsg'

Designed to escape sudden chemical, biological, radiological, or nuclear hazards

  • Translucent hood with one-piece large bonded lens offers secure protection and a wide field of view
  • Canister is permanently bonded to the respirator for a secure connection
  • Rubber neck seal provides a comfortable and reliable seal
  • Elastic hair tie allows for the securing of hair before donning the respirator
  • Nose cup helps reduce lens fogging and carbon dioxide buildup
  • Hexagonal storage case is strong, lightweight, and easily accessible
  • 15 minute air supply

Standard

  • Color-coded labels on respirator
  • Tactical
    • Black/gray labels on respirator

    Guards against HD vapor, GB, Ammonia, Cyanogen chloride, Cyclohexane, Formaldehyde, Hydrogen cyanide, Hydrogen sulfide, Nitrogen dioxide, Phosgene, Phosphine, sulfur dioxide

    Certifications

    NIOSH - 10052317

    Specifications

    Certifications/Compliance NIOSH
    Product Type Tactical Escape Respirator
    Duration 15 min.
    Cycle Time 15 min.
    For Use With (Application) CBRN incidents, civil disturbances, clandestine labs
    Includes Bonded lens and cbrn canister, high stretch rubber neck seal, nose cup
    Model 10066789
    Size Large
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.