Learn More
MSA™ Safe Escape CBRN Training Hoods
Catalog No. 19161119 Shop All MSA Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-161-119 Large
19-161-117 Small
19-161-118 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-161-119 Supplier MSA™ Supplier No. 10066786
Guards against HD vapor, GB, Ammonia, Cyanogen chloride, Cyclohexane, Formaldehyde, Hydrogen cyanide, Hydrogen sulfide, Nitrogen dioxide, Phosgene, Phosphine, sulfur dioxide
Certifications
NIOSH - 10052317
Specifications
Certifications/Compliance NIOSH
Product Type Training Hood
For Use With (Application) CBRN incidents, civil disturbances, clandestine labs
For Use With (Equipment) Emergency escape respirator
Model 10066786
Size Large