Learn More
Description
The one-piece wraparound glasses protect against impact hazards and/or flying particles, dust, sparks and glare.
- Frameless design with soft nose pads
- Horizontal temple adjustment
- Wrap-style single lens
- Tuff Stuff™ anti-scratch coating
- Available in clear or gray lenses
- For painting and food processing applications
Certifications
CSA Z94.3 certified
Compliance
ANSI/ISEA Z87.1-2010 compliant
Specifications
Specifications
| Lens Coating | Scratch-resistant |
| Lens Tint | Gray |
| Product Type | Protective Eyewear |
| Temple Style | Adjustable |
| Material | Polycarbonate |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |
| UV Protection | U6 |
| Frame Type | Frameless |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.