Learn More
MSA™ Skullgard™ Protective Caps
Catalog No. 19133741 Shop All MSA Products
Change view
Click to view available options
Size:
Large
Catalog No. Size
19-133-741 Large
1 options
Catalog No. 19-133-741 Supplier MSA™ Supplier No. 475405
Only null left
Add to Cart
Add to Cart
Capacity of withstanding radiant heat loads up to 176.6°C (350°F)
  • Feature heavy-duty construction for use in steel mills other heavy industries
  • Smooth crown for easy use of welding equipment
  • Used in food processes, asbestos abatement, painting
Compliance
Meet ANSI/ISEA Z89.1-2009 (Class G)
Specifications
Size Large
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.