Learn More
MSA™ Ultra Elite Spectacle Kit for Self-Contained Breathing Apparatus (SCBA)
Catalog No. 189993536 Shop All MSA Products
Change view
Click to view available options
Includes:
Center wire support, rubber guide, pair of S-7 shaped metal frame spectacles
Side wire support, rubber guide, pair of S-7 shaped metal frame spectacles
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
18-999-3536 Side wire support, rubber guide, pair of S-7 shaped metal frame spectacles
18-999-3522 Center wire support, rubber guide, pair of S-7 shaped metal frame spectacles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-999-3536 Supplier MSA™ Supplier No. 804638
Add to Cart
Add to Cart
Includes
Note: These do not include lenses - just wire support and frames.
Spectacle kits allow full facepieces to accommodate eyeglasses.
Kits allow Ultra Elite™ facepiece to accommodate corrective lenses. Include wire support and rubber guide for easy custom adjustment and S-7 shaped metal spectacles frame for use with 44mm universal bridge corrective lenses. Choose from either side-wire or center support kit.
Specifications
Applications Firefighting
For Use With (Equipment) Ultra-Elite Facepiece
Includes Side wire support, rubber guide, pair of S-7 shaped metal frame spectacles
Model 804638
Product Type Spectacle/Lens Kit