Learn More
MSA™ Standard Body Belts
Catalog No. 19015178 Shop All MSA Products
Change view
Click to view available options
Size:
Small
Medium
Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-015178 Small
19-015181 Large
19-815-449 -
19-815-450 Medium
19-046-113 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Must never be used for fall arrest
Catalog No. 19-015178 Supplier MSA™ Supplier No. 415277
Add to Cart
Add to Cart
Must never be used for fall arrest
Belts to protect back while lifting
  • Fits 78.74cm (32 in.) to 104.14cm (41 in.) waist
  • Offered in three differest sizes
Restraint, travel restriction, work positioning
Specifications
Size Small
D-Ring Location Two
For Use With (Application) Used for restraint, travel restriction or work positioning; Not for fall arrest
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.