Learn More
MSA™ Sure-Line™ Horizontal Lifelines
Catalog No. 191600215 Shop All MSA Products
Change view
Click to view available options
Steel
Catalog No.
19-160-0215 Steel
1 options
Catalog No. 19-160-0215 Supplier MSA™ Supplier No. SHL2009060
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
For personal fall arrest on horizontal plane
  • For two workers
  • No multiple anchorage points required
  • 60 ft.
  • Galvanized cable
  • Integral shock absorber with clamp
  • Made of steel
Specifications
Product Type Sure-Line™ horizontal lifeline
For Use With (Equipment) Personal fall arrest on horizontal plane
Certifications/Compliance OSHA, ANSI A10.32
Includes Turnbuckle, Galvanized cable, integral shock absorber with clamp
Length (Metric) 18.3 m
Length (English) 60 ft.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.