Learn More
MSA™ Tee Blocks and Pigtails
Catalog No. 18139728 Shop All MSA Products
Change view
Click to view available options
Connection Type:
Air pigtail
Air tee block
Material:
Brass
Tubular Copper
Product Type:
Pigtail
Tee Block
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type Material Product Type
18-139-728 Air pigtail Tubular Copper Pigtail
18-139-727 Air tee block Brass Tee Block
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-139-728 Supplier MSA™ Supplier No. 68851
Add to Cart
Add to Cart
Tee blocks and pigtails are used to connect a bank of cylinders together.
Used to connect a bank of cylinders together
  • Coupler tees make a connection between the supply cylinder valve and the pigtail
  • Outlet connection are constructed of brass, while the pigtails joining the supply cylinders are made of tubular copper
  • 27 in. long air pigtail
Specifications
Connection Type Air pigtail
For Use With (Equipment) Compressed Airline Respirator
Material Tubular Copper
Model 68851
Pressure 3000 psi
Overall Length (English) 27 in.
Overall Length (Metric) 68.58 cm
Product Type Pigtail