Learn More
MSA™ Thermalgard™ Protective Caps
Catalog No. 19035054 Shop All MSA Products
Change view
Click to view available options
Color:
Bright Orange
Green
Suspension Type:
1-Touch
Fas-Trac Ratchet
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Suspension Type
19-035-054 Green 1-Touch
18-999-3813 Bright Orange 1-Touch
18-999-3816 Green Fas-Trac Ratchet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-035-054 Supplier MSA™ Supplier No. 487399
Add to Cart
Add to Cart
Lightweight, high-strength helmet with elevated temperature capability
This slotted cap is intended for use where workers may need a helmet with elevated temperature capability.
  • Injection-molded, rivetless shell of glass-reinforced nylon has beaded edge for improved rigidity
  • Lightweight, high-strength helmets for use in elevated temperatures; withstands radiant heat loads up to, but not exceeding, 350°F
  • Slots in cap allow for mounting of MSA hearing protection, welding helmets, and V-Gard System face protection products, as well as other MSA helmet accessories
Compliance
Third-party certified by SEI to meet the Type I, Class G requirements of ANSI/ISEA Z98.1 and CSA Z94.1.
Specifications
Certifications/Compliance Meets ANSI Z89.1-2003; Type I; Class G and CSA Z94.1-2005 (Class G).
Color Green
Suspension Type 1-Touch
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.