Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Twin-Cartridge Facepiece Adapter Assembly, Use with Advantage 4000 Facepieces

Catalog No. 19716758 Shop All MSA Products
Change view
Click to view available options
Cartridge adapter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-716-758 Cartridge adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-716-758 Supplier MSA™ Supplier No. 10065328
Add to Cart
Add to Cart

Twin-cartridge adapter for use with Advantage 4000 series respirators.

  • For use with Advantage 4000 respirator
  • Twin cartridge adapter MSA 10065328

Specifications

Color Black
Product Type Adapter
Size One Size
For Use With (Equipment) Advantage 4000 Full Facepieces
Model 10065328
Dimensions (L x W x H) 5.5 x 3.5 x 3.0 in. (14.0 x 8.9 x 7.6 cm)
Weight (English) 0.5 lb.
Weight (Metric) 227 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.